missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH11X Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its Y homolog, despite divergence leading to coding region changes, is the most closely related cadherin family member. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Disruption of this gene may be associated with developmental dyslexia. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | PCDH11X |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PCDH11X |
| Gene Accession No. | 0 |
| Gene Alias | KIAA1326; PCDH11; PCDH11X; PCDH11Y; PCDHX; PCDH-X; PCDHX11; PPP1R119; protein phosphatase 1, regulatory subunit 119; protocadherin 11 X-linked; protocadherin 11 Y-linked; protocadherin 11X; protocadherin on the X chromosome; protocadherin X-linked 11; Protocadherin-11; Protocadherin-11 X-linked; protocadherin-S; RGD1562864 |
| Gene Symbols | PCDH11X |
| Show More |