missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCDH10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531042 Supplier Invitrogen™ Supplier No. PA531042
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (95%), Chicken (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains.

Specifications

Antigen PCDH10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PCDH10
Gene Accession No. Q9P2E7
Gene Alias 6430521D13Rik; 6430703F07Rik; KIAA1400; LOW QUALITY PROTEIN: protocadherin-10; mKIAA1400; OL-pc; OL-PCDH; OL-protocadherin; ortholog of OL-pcdh; PCDH10; PCDH19; protocadherin 10; protocadherin-10; RGD1565811
Gene Symbols PCDH10
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human PCDH10. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less