missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCB Congeners - Loamy Sand, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 39635-31-9 |
| Type | Certified Reference Material |
| Quantity | 50 g |
| Grade | Certified Reference Material |