Learn More
MilliporeSigma™ Calbiochem™ PBS-TWEEN™ Tablets
Dissolve one tablet in 1 liter of deionized water to yield 140 mM NaCl, 3 mM KCl, 0.05% TWEEN™ 20 Detergent, 10 mM phosphate buffer, pH 7.4 at 25°C
Supplier: MilliporeSigma™ 5246531EA
Description
Following reconstitution, store in the refrigerator (4°C) or filter sterilize and store at room temperature (20°C)Greener Choice Claims
- Environmental benefits include:
- Recyclable
- Certifications and Seals of Approval
Specifications
10 Tablets | |
Room Temperature |
Promotions
-
MilliporeSigma
,
Merck Emd Millipore
,
Technidata
Get More MilliporeSigma Products with Purchase
Spend up to $1,500 on MilliporeSigma products and get a matching dollar amount to spend on your next purchase.Expires: 06/30/26 Redemption Only