Learn More
Invitrogen™ Human PBK (TOPK), His Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PV4285
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
PBK (TOPK) is a serine/threonine kinase that is related to the dual specific mitogen-activated protein kinase (MAPKK). The kinase is up-regulated in a variety of hematological malignancies, participates in activating lymphoid cells, and supports testicular function (spermatogenesis). Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.Specifications
| Q96KB5 | |
| Kinase Assay | |
| 55872 | |
| Human PBK (TOPK), His Tag | |
| RUO - research use only | |
| PBK | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Insect cells |
| See Label | |
| 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 | |
| 41 kDa | |
| 1 mg | |
| 2810434B10Rik; AW538537; Cancer/testis antigen 84; CT84; D14Ertd732e; epididymis luminal protein 164; HEL164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PBK; PDZ binding kinase; PDZ-binding kinase; serine/threonine protein kinase; spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase; TOPK | |
| TOPK Kinase Family | |
| Human | |
| His-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Liquid |