missing translation for 'onlineSavingsMsg'
Learn More

Paxilline, 97+%, Thermo Scientific Chemicals

Catalog No. aaj61341lb0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61341LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61341LB0 Supplier Thermo Scientific Chemicals Supplier No. J61341LB0
Only null left

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57186-25-1
Molecular Formula C27H33NO4
Molecular Weight (g/mol) 435.564
MDL Number MFCD00083464
InChI Key ACNHBCIZLNNLRS-UBGQALKQSA-N
Synonym paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one
PubChem CID 105008
ChEBI CHEBI:34907
SMILES CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O

Specifications

Color White to Yellow
Quantity 5 mg
UN Number UN2811
Solubility Information Soluble in DMSO,acetone or chloroform.
Formula Weight 435.56
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Paxilline
Hazard Category H301+H311-H315-H318-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber DJ2830000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only