accessibility menu, dialog, popup

UserName

CAS RN 57186-25-1

IUPAC Name: (1S,2R,5S,7R,11S,14S)-11-hydroxy-7-(2-hydroxypropan-2-yl)-1,2-dimethyl-6-oxa-23-azahexacyclo[12.10.0.0²,¹¹.0⁵,¹⁰.0¹⁶,²⁴.0¹⁷,²²]tetracosa-9,16(24),17,19,21-pentaen-8-one
Synonyms: paxilline
Molecular Weight (g/mol): 435.56
Molecular Formula: C27H33NO4
InChi Key: ACNHBCIZLNNLRS-UBGQALKQSA-N
SMILES: CC(C)(O)[C@H]1O[C@H]2CC[C@]3(C)[C@]4(C)[C@H](CC5=C4NC4=CC=CC=C54)CC[C@@]3(O)C2=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results