missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Paxillin Recombinant Rabbit Monoclonal Antibody (23GB2690)
Rabbit Recombinant Monoclonal Antibody
$656.94 - $1779.35
Specifications
| Antigen | Paxillin |
|---|---|
| Clone | 23GB2690 |
| Concentration | 0.12 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
Paxillin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.Specifications
| Paxillin | |
| 0.12 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49023, Q66H76, Q8VI36 | |
| 19303, 360820, 5829 | |
| A synthesized peptide derived from human Paxillin (1-39AA). | |
| Antibody |
| 23GB2690 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PXN | |
| adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 | |
| PXN | |
| Primary | |
| Affinity chromatography |