missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAX9 Recombinant Rabbit Monoclonal Antibody (3C2U5)
GREENER_CHOICE

Catalog No. PIMA537975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537975 Supplier Invitrogen™ Supplier No. MA537975
Only null left

Rabbit Recombinant Monoclonal Antibody

Positive Samples: PC-3, Mouse stomach, Mouse thymus, Rat lung, Rat stomach Immunogen sequence: MEPAFGEVNQ LGGVFVNGRP LPNAIRLRIV ELAQLGIRPC DISRQLRVSH GCVSKILARY NETGSILPGA IGGSKPRVTT PTVVKHIRTY KQRDPGIFAW.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements.

Specifications

Antigen PAX9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 3C2U5
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene pax9
Gene Accession No. P47242, P55771, Q2L4T2
Gene Alias EMBL:BAE79272.1}; etID309776.17; paired box 9; paired box gene 9; paired box homeotic gene 9; paired box protein Pax-9; paired domain gene 9; Pax9; Pax-9; pax9 {ECO:0000312; pax9 protein; Pax9a; Pax9b; STHAG3
Gene Symbols pax9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PAX9 (P55771).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18511, 362741, 5083
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less