missing translation for 'onlineSavingsMsg'
Learn More

Patulin Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018028
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018028 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018028 Supplier Merck Emd Millipore Supplier No. 341272ML

Patulin is a mycotoxin produced by several mold species. It has been found in fruit, especially apples, fruit juice, cider and stewed fruits. Because of its highly toxic properties, this compound has been the focus of many studies.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 149-29-1
Linear Formula C7H6O4
MDL Number MFCD00005858
Synonym 4-Hydroxy-4 H-furo[3,2-c]pyran-2(6 H)-one; PAT
Type Analytical Standard
Quantity 2 mL
Formula Weight 154.12
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile