Learn More
Particulate Matter in Impinger Solution, Proficiency Testing Material, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Supplier:  Merck Emd Millipore PEA1906250ML
Catalog No. 111004502
Only null left
Explore available promotions
Description
Description
This PT is packaged as a 250 mL whole-volume.The sample matrix is water. This PT is suitable for EPA Methods 5, 5A, 5B, 5D, and 5F.Sample preparationMix by inverting the container 10 times. Let the container sit for 10 minutes and invert again 10 times.When transferring to another container, repetitively mix and pour so as not to allow solids to settle before transfer is complete.The sample is now ready for immediate analysis.
Specifications
Specifications
250 mL
Particulate Matter 25-500 mg/L
Product Suggestions
Product Suggestions