missing translation for 'onlineSavingsMsg'
Learn More

PARP3 Antibody, ProSci

Catalog No. 16825411
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16825411 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16825411 Supplier ProSci Llc Supplier No. 467530100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Goat Polyclonal Antibody

PARP3 Polyclonal Antibody specifically detects PARP3 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen PARP3
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 500 ug/mL
Conjugate Unconjugated
Dilution Peptide ELISA: antibody detection limit dilution 1:2000.Western Blot:Preliminary experiments in lysates of cell lines Daudi, HeLa and HepG2 gave no specific signal but low background (at antibody concentration up to 1ug/mL).Immunohistochemistry:Paraffin embedded Human Liver and Kidney. Recommended concentration: 3.75ug/mL.
Formulation Supplied at 0.5 mg/mL in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
Gene Accession No. NP_001003931.2,NP_001003935.2
Gene Alias PARP3, poly (ADP-ribose) polymerase family, member 3, ADPRT3, ADPRTL2, ADPRTL3, IRT1, hPARP-3, pADPRT-3, ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase)-like 2, ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase)-like 3, NAD+ ADP-ribosyltransferase 3, poly(ADP-ribose) polymerase 3, poly(ADP-ribose) synthetase-3
Host Species Goat
Purification Method Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10039
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001003931.2; NP_001003935.2).
Target Species Human
Content And Storage Aliquot and store at -20°C. Minimize freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Apoptosis
Show More Show Less