missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals PARP Inhibitor VIII, PJ34
Description
A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
Specifications
Specifications
| Quantity | 5 mg |
| Description | White |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase |
| Formula Weight | 331.80g/mol |
| Sensitivity | Light Sensitive and Hygroscopic |
| Product Type | Signal Transduction Reagent |
| Form | Solid |
| InChI Key | RURAZZMDMNRXMI-UHFFFAOYSA-N |
| SMILES | Cl.CN(C)CC(=O)NC1=CC=C2NC(=O)C3=CC=CC=C3C2=C1 |
| Show More |
Safety and Handling
| Recommended Storage | Store at -20°C |
RUO – Research Use Only