missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paromomycin Sulfate 94.0+%, TCI America™
$100.67 - $298.25
Chemical Identifiers
| CAS | 1263-89-4 |
|---|---|
| Molecular Formula | C23H47N5O18S |
| Molecular Weight (g/mol) | 713.706 |
| MDL Number | MFCD00079278 |
| InChI Key | LJRDOKAZOAKLDU-JXDXLELXSA-N |
| PubChem CID | 91972180 |
| IUPAC Name | (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid |
| SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O |
Chemical Identifiers
| 1263-89-4 | |
| 713.706 | |
| LJRDOKAZOAKLDU-JXDXLELXSA-N | |
| (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid |
| C23H47N5O18S | |
| MFCD00079278 | |
| 91972180 | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O |
Specifications
| Paromomycin Sulfate | |
| 1263-89-4 | |
| 1 g | |
| MFCD00079278 | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O | |
| 713.706 | |
| ≥94.0% (HPLC) |
| Crystalline Powder | |
| White/Yellow | |
| C23H47N5O18S | |
| LJRDOKAZOAKLDU-JXDXLELXSA-N | |
| (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid | |
| 91972180 | |
| Paromomycin Sulfate |
Safety and Handling
RTECSNumber : WK2320000
TSCA : No