missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Parainfluenza Virus Type 3 Monoclonal Antibody (F96H), FITC

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17319
Description
The MA1-7319 antibody reacts with Parainfluenza virus 3 in viral and infected tissue samples. MA1-7319 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7319 antibody was raised against Parainfluenza Type 3 antigen.
Chemical Identifiers
| Parainfluenza Virus Type 3 | |
| 0.1 mg/mL | |
| ELISA, Immunocytochemistry | |
| FITC | |
| Mouse | |
| RUO | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Parainfluenza Type 3 antigen. | |
| Antibody |
| F96H | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C. Store in the dark. | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Virus | |
| canine parainfluenza virus; hPIV-3; PIV-3 | |
| Primary | |
| Protein A |
Specifications
| Parainfluenza Virus Type 3 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Immunocytochemistry | |
| F96H | |
| FITC | |
| canine parainfluenza virus; hPIV-3; PIV-3 | |
| Parainfluenza Type 3 antigen. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C. Store in the dark. | |
| Liquid |