Learn More
Thermo Scientific Chemicals Paraformaldehyde, 8% w/v aq. soln., methanol free
Solution is micro-filtered and supplied in ampules. | CAS: 50-00-0 | CH2O | 30.03 g/mol
$71.38 - $161.21
Chemical Identifiers
| CAS | 50-00-0 |
|---|---|
| Molecular Formula | CH2O |
| Molecular Weight (g/mol) | 30.03 |
| MDL Number | MFCD00133991 |
| InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
| PubChem CID | 712 |
| ChEBI | CHEBI:16842 |
| IUPAC Name | formaldehyde |
| SMILES | C=O |
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Price | Quantity & Availability | |||||
|
AA473479L
|
Thermo Scientific Chemicals
0473479L |
2 x 10 mL |
Case of 2 Each for $71.38
|
|
|||||
|
AA473479M
|
Thermo Scientific Chemicals
0473479M |
10 x 10 mL |
Case of 10 Each for $161.21
|
|
|||||
Description
Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.Chemical Identifiers
| 50-00-0 | |
| 30.03 | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| CHEBI:16842 | |
| C=O |
| CH2O | |
| MFCD00133991 | |
| 712 | |
| formaldehyde |
Specifications
| 50-00-0 | |
| (CH2O)n | |
| 102769 | |
| Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol. | |
| C=O | |
| 30.03 | |
| CHEBI:16842 | |
| Liquid |
| CH2O | |
| MFCD00133991 | |
| 14,7025 | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| formaldehyde | |
| 712 | |
| 8% w/v aq. soln. | |
| Paraformaldehyde |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H332-H314-H317-H335-H341-H350
EINECSNumber : 200-001-8
RTECSNumber : RV0540000
TSCA : Yes
storageNote1 : Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only