missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 4% w/v aq. soln., methanol free

Solution is micro-filtered and supplied in ampules. | CAS: 50-00-0 | CH2O | 30.03 g/mol

Supplier:  Thermo Scientific Chemicals 0473929M

 View more versions of this product

Catalog No. AA473929M


Only null left

Description

Description

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to fluorescence studies.
Specifications

Specifications

Paraformaldehyde
CH2O
MFCD00133991
102769
Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol.
C=O
30.03
CHEBI:16842
Liquid
50-00-0
(CH2O)n
10 x 10 mL
14,7025
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
4% w/v aq. soln.

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
CHEBI:16842
C=O
CH2O
MFCD00133991
712
formaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P271-P272-P280g-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P333+P313-P362-P501c

H315-H317-H318-H335-H341-H350

EINECSNumber : 200-001-8

RTECSNumber : RV0540000

TSCA : Yes

storageNote1 : Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only