missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, 32% w/v aq. soln., methanol free

Solution is micro-filtered and supplied in ampules. | CAS: 50-00-0 | CH2O | 30.03 g/mol

$111.42 - $228.07

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA473779L
View Documents Promotion Details
N/A 2 x 10 mL
Case of 2 Each for $111.42
Only null left
 
AA473779M
View Documents Promotion Details
N/A 10 x 10 mL
Case of 10 Each for $228.07
Only null left
 
Description

Description

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
Specifications

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
CHEBI:16842
C=O
CH2O
MFCD00133991
712
formaldehyde

Specifications

50-00-0
(CH2O)n
102769
Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol.
C=O
30.03
CHEBI:16842
Liquid
CH2O
MFCD00133991
14,7025
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
32% w/v aq. soln.
Paraformaldehyde
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H332-H311-H314-H317-H335-H341-H350

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 200-001-8

RTECSNumber : RV0540000

TSCA : Yes

storageNote1 : Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only