missing translation for 'onlineSavingsMsg'
Learn More

Paraben internal standard mix Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111016612
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016612 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016612 Supplier Merck Emd Millipore Supplier No. 321245ML
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 67-64-1
Flash Point -17.0°C - Closed Cup
Type Analytical Standard
Quantity 5 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in acetone