missing translation for 'onlineSavingsMsg'
Learn More

Parabanic Acid 98.0+%, TCI America™

Catalog No. p-7134478
Change view
Click to view available options
Quantity:
1 g
25 g
Catalog No. Quantity
P01951G 1 g
P019525G 25 g
2 options

Catalog No. P01951G

Supplier: TCI America P01951G

Only null left

Chemical Identifiers

CAS 120-89-8
Molecular Formula C3H2N2O3
Molecular Weight (g/mol) 114.06
MDL Number MFCD00014493
InChI Key ZFLIKDUSUDBGCD-UHFFFAOYSA-N
PubChem CID 67126
ChEBI CHEBI:74661
IUPAC Name imidazolidine-2,4,5-trione
SMILES O=C1NC(=O)C(=O)N1

Specifications

Melting Point 249°C
Color White
Quantity 1 g
Synonym N,N′C-Oxalylurea
Formula Weight 114.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Parabanic Acid