missing translation for 'onlineSavingsMsg'
Learn More

Parabanic Acid 98.0+%, TCI America™

Catalog No. P019525G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P019525G 25 g
P01951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P019525G Supplier TCI America Supplier No. P019525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-89-8
Molecular Formula C3H2N2O3
Molecular Weight (g/mol) 114.06
MDL Number MFCD00014493
InChI Key ZFLIKDUSUDBGCD-UHFFFAOYSA-N
PubChem CID 67126
ChEBI CHEBI:74661
IUPAC Name imidazolidine-2,4,5-trione
SMILES O=C1NC(=O)C(=O)N1

Specifications

Melting Point 249°C
Color White
Quantity 25 g
Synonym N,N′-Oxalylurea
Formula Weight 114.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Parabanic Acid