missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAPPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149789
Description
PAPPA Polyclonal Antibody for IHC (P), Western Blot
Pregnancy-associated plasma protein-A (PAPP-A, Insulin-like growth factor binding protein-4 protease, PAPP-A-proMBP complex) is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down's syndrome (trisomy 21).Specifications
| PAPPA | |
| Polyclonal | |
| Unconjugated | |
| PAPPA | |
| 8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18491, 5069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q13219, Q8R4K8 | |
| PAPPA | |
| Recombinant Pregnancy Associated Plasma Protein A (PAPPA) (Phe999-Asn1272). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |