Learn More
Motic Panthera C2 Microscope and Camera Bundle
The Panthera C2 is prepared for Phase Contrast with slider or turret condenser, Polarization Contrast, and Darkfield. It is even possible to integrate a ready-to-go LED fluorescence module, allowing for advanced methods without problems associated with mercury bulb usage.
Supplier:  Motic 1100403300222
 View more versions of this product
Catalog No. 01-257-486
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Swiveling, 25° inclined Siedentopf head
  • Nosepiece with light tracer (Intelli Light Management)
  • LED light indicator
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built-in, low position coaxial rackless stage with right hand control
  • Focusable Abbe condenser with 0.9 numerical aperature
  • Iris diaphragm and slot
  • Diopter Adjustment: ±5
Specifications
Specifications
BTI10 Camera Bundle
CCIS Plan UC Objectives 4X, 10X, 40X S, 100X S-oil
Trinocular
LED
10X
Panthera C2 Trinocular
Moticam BTI10
WF 10X/22 mm
Product Suggestions
Product Suggestions