missing translation for 'onlineSavingsMsg'
Learn More

Medisca Panthenol USP

Catalog No. 14186118
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186118 1 kg
14186097 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14186118 Supplier Medisca Supplier No. 148109
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Panthenol USP is a white to creamy white, crystalline powder having a slight, characteristic odor. It is hygroscopic.

Panthenol USP is a white to creamy white, crystalline powder having a slight, characteristic odor. It is hygroscopic.

Alternative name(s): DL-Panthenol, DL-Pantothenyl alcohol

Specifications

Chemical Name or Material Panthenol USP
Packaging Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck
CAS 16485-10-2
Type Drug Standard
Molecular Weight (g/mol) 205.25
Quantity 1 kg
Percent Purity 99 to 102 %
Grade USP