missing translation for 'onlineSavingsMsg'
Learn More

PAN Indicator, 0.3% in Ethanol, Certified, LabChem™

Supplier:  LabChem LC180901

 View more versions of this product

Catalog No. LC180901


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

PAN Indicator
89.7,5,5,0.3
C15H11N3O
Soluble in water
249.27
Amber Glass
Carbon monoxide; Carbon dioxide; May release flammable gases
500 mL
0.3% in Ethanol
67-56-1 , 67-63-0 , 85-85-8
UN1170
Passes Test
Certified
Passes Test
Orange to Red
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions