missing translation for 'onlineSavingsMsg'
Learn More

Pamoic Acid 96.0+%, TCI America™

Catalog No. p076125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P076125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P076125G Supplier TCI America Supplier No. P076125G
Only null left

Chemical Identifiers

CAS 130-85-8
Molecular Formula C23H16O6
Molecular Weight (g/mol) 388.38
MDL Number MFCD00004079
InChI Key WLJNZVDCPSBLRP-UHFFFAOYSA-N
Synonym pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure
PubChem CID 8546
ChEBI CHEBI:50186
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1

Specifications

Formula Weight 388.38
Percent Purity ≥96.0% (HPLC)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Pamoic Acid
EINECSNumber (4)-0462
RTECSNumber QL2180000
TSCA Yes