missing translation for 'onlineSavingsMsg'
Learn More

Pamoic acid, 99%

Catalog No. aaa1520718
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1520718 50 g
AAA1520730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1520718 Supplier Thermo Scientific Chemicals Supplier No. A1520718
Only null left

CAS: 130-85-8 | C23H16O6 | 388.38 g/mol

Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

Solubility
Soluble in nitrobenzene, pyridine

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 130-85-8
Molecular Formula C23H16O6
Molecular Weight (g/mol) 388.38
MDL Number MFCD00004079
InChI Key WLJNZVDCPSBLRP-UHFFFAOYSA-N
Synonym pamoic acid, embonic acid, 4,4'-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7rrq8qz38n, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy, 7rrq8qz38n, pamoic acid 98+% hplc, 4,4'-methylen-bis-3-hydroxy-2-naphthoesaeure
PubChem CID 8546
ChEBI CHEBI:50186
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1

Specifications

Melting Point >300°C
Assay Percent Range 99%
Beilstein 901319
Merck Index 14,7005
Solubility Information Soluble in nitrobenzene, pyridine
Formula Weight 388.38
Percent Purity 99%
Quantity 50 g
Chemical Name or Material Pamoic Acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-998-0
RTECSNumber QL2180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only