missing translation for 'onlineSavingsMsg'
Learn More

Pamabrom 98.0+%, TCI America™

Catalog No. P23591G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
P23591G 1 g
P23595G 5 g
2 options

Catalog No. P23591G

Supplier: TCI America P23591G

Only null left

Chemical Identifiers

CAS 606-04-2
Molecular Formula C11H18BrN5O3
Molecular Weight (g/mol) 348.201
MDL Number MFCD00867328
InChI Key ATOTUUBRFJHZQG-UHFFFAOYSA-N
Synonym 8-Bromotheophylline 2-Amino-2-methyl-1-propanol
PubChem CID 11806
IUPAC Name 2-amino-2-methylpropan-1-ol;8-bromo-1,3-dimethyl-7H-purine-2,6-dione
SMILES CC(C)(CO)N.CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br

Specifications

Melting Point 295°C
Color White
Quantity 1 g
Formula Weight 348.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pamabrom