missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitoyl-L-carnitine-(N-methyl-d3), ≥97.0% (TLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 202480-73-7 |
| Linear Formula | C23D3H42 NO4 |
| MDL Number | MFCD01073471 |
| Synonym | (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt; L-Carnitine-(methyl-d3) hexadecanoyl ester; Hexadecanoyl-L-carnitine-(N-methyl-d3) |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 402.63 |
| Percent Purity | ≥97.0% (TLC) |
| Grade | Analytical Standard |