missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitoyl-L-carnitine, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 2364-67-2 |
| Linear Formula | C23H45 NO4 |
| Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt; L-Carnitine hexadecanoyl ester; C16-Carnitine; Hexadecanoyl-L-carnitine |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 399.61 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |