missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitoyl Chloride 97.0+%, TCI America™
$42.87 - $177.08
Chemical Identifiers
| CAS | 112-67-4 |
|---|---|
| Molecular Formula | C16H31ClO |
| Molecular Weight (g/mol) | 274.873 |
| MDL Number | MFCD00000742 |
| InChI Key | ARBOVOVUTSQWSS-UHFFFAOYSA-N |
| Synonym | palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride |
| PubChem CID | 8206 |
| IUPAC Name | hexadecanoyl chloride |
| SMILES | CCCCCCCCCCCCCCCC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P000925ML
|
TCI America
P000925ML |
25 mL |
Each for $42.87
|
|
|||||
|
P0009500ML
|
TCI America
P0009500ML |
500 mL |
Each for $177.08
|
|
|||||
Chemical Identifiers
| 112-67-4 | |
| 274.873 | |
| ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
| 8206 | |
| CCCCCCCCCCCCCCCC(=O)Cl |
| C16H31ClO | |
| MFCD00000742 | |
| palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
| hexadecanoyl chloride |
Specifications
| 112-67-4 | |
| 200°C | |
| MFCD00000742 | |
| 3265 | |
| ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
| hexadecanoyl chloride | |
| 8206 | |
| ≥97.0% (T) | |
| Palmitoyl Chloride |
| Yellow | |
| C16H31ClO | |
| 25 mL | |
| palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
| CCCCCCCCCCCCCCCC(=O)Cl | |
| 274.873 | |
| 274.87 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes
Recommended Storage : Refrigerator