missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl Chloride 97.0+%, TCI America™

Catalog No. P000925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P000925ML 25 mL
P0009500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P000925ML Supplier TCI America Supplier No. P000925ML

Chemical Identifiers

CAS 112-67-4
Molecular Formula C16H31ClO
Molecular Weight (g/mol) 274.873
MDL Number MFCD00000742
InChI Key ARBOVOVUTSQWSS-UHFFFAOYSA-N
Synonym palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
PubChem CID 8206
IUPAC Name hexadecanoyl chloride
SMILES CCCCCCCCCCCCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 mL
UN Number 3265
Formula Weight 274.87
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Palmitoyl Chloride
EINECSNumber (2)-0632
TSCA Yes
Recommended Storage Refrigerator