Learn More
Palmitoyl chloride, 98%
CAS: 112-67-4 | C16H31ClO | 274.88 g/mol
Supplier: Thermo Scientific Chemicals 304290025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Palmitoyl chloride | |
| 112-67-4 | |
| 9.0°C to 14.0°C | |
| 199.0°C (2.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4510 to 1.4530 | |
| MFCD00000742 | |
| 02, 374 | |
| palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
| CCCCCCCCCCCCCCCC(=O)Cl | |
| 274.873 | |
| 274.88 | |
| Liquid |
| 98% | |
| 1% max. (as palmitic acid) | |
| 0.9000g/mL | |
| 160°C | |
| 98% | |
| C16H31ClO | |
| CH3(CH2)14COCl | |
| 2.5 L | |
| 0.9 | |
| ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
| hexadecanoyl chloride | |
| 8206 | |
| 98% |
Chemical Identifiers
| 112-67-4 | |
| ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
| 8206 | |
| CCCCCCCCCCCCCCCC(=O)Cl |
| MFCD00000742 | |
| palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
| hexadecanoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.
Danger
EINECSNumber : 203-996-7
TSCA : TSCA
RUO – Research Use Only