Learn More
Thermo Scientific Chemicals Palmitoleic acid, 99%
CAS: 373-49-9 | C16H30O2 | 254.41 g/mol
$155.86 - $356.32
Chemical Identifiers
| CAS | 373-49-9 |
|---|---|
| Molecular Formula | C16H30O2 |
| Molecular Weight (g/mol) | 254.41 |
| MDL Number | MFCD00004437 |
| InChI Key | SECPZKHBENQXJG-FPLPWBNLSA-N |
| Synonym | palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid |
| PubChem CID | 445638 |
| ChEBI | CHEBI:28716 |
| IUPAC Name | (Z)-hexadec-9-enoic acid |
| SMILES | CCCCCC\C=C/CCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC376912500
|
Thermo Scientific Chemicals
376912500 |
250 mg | Glass bottle |
Each for $155.86
|
|
||||
|
AC376910010
|
Thermo Scientific Chemicals
376910010 |
1 g | Glass bottle |
Each for $356.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 373-49-9 | |
| 254.41 | |
| SECPZKHBENQXJG-FPLPWBNLSA-N | |
| 445638 | |
| (Z)-hexadec-9-enoic acid |
| C16H30O2 | |
| MFCD00004437 | |
| palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid | |
| CHEBI:28716 | |
| CCCCCC\C=C/CCCCCCCC(O)=O |
Specifications
| 373-49-9 | |
| 0.8940g/mL | |
| 62°C | |
| 98.5% min. (GC) | |
| C16H30O2 | |
| MFCD00004437 | |
| 0.894 | |
| Solubility in water: insoluble | |
| CCCCCC\C=C/CCCCCCCC(O)=O | |
| 254.41 | |
| CHEBI:28716 | |
| 99% | |
| Palmitoleic acid |
| 0.5°C | |
| 162.0°C (0.6 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4570 to 1.4590 | |
| 250 mg | |
| palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid | |
| SECPZKHBENQXJG-FPLPWBNLSA-N | |
| (Z)-hexadec-9-enoic acid | |
| 445638 | |
| 254.41 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 206-765-9
RUO – Research Use Only