missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Palmitoleic acid, 99%

CAS: 373-49-9 | C16H30O2 | 254.41 g/mol

Supplier:  Thermo Scientific Chemicals 376910010

 View more versions of this product

Catalog No. AC376910010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Palmitoleic acid
0.5°C
162.0°C (0.6 mmHg)
Authentic
Glass bottle
1.4570 to 1.4590
1 g
palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
SECPZKHBENQXJG-FPLPWBNLSA-N
(Z)-hexadec-9-enoic acid
445638
254.41
Liquid
373-49-9
0.8940g/mL
62°C
98.5% min. (GC)
C16H30O2
MFCD00004437
0.894
Solubility in water: insoluble
CCCCCC\C=C/CCCCCCCC(O)=O
254.41
CHEBI:28716
99%

Chemical Identifiers

373-49-9
254.41
SECPZKHBENQXJG-FPLPWBNLSA-N
445638
(Z)-hexadec-9-enoic acid
C16H30O2
MFCD00004437
palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
CHEBI:28716
CCCCCC\C=C/CCCCCCCC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 206-765-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only