missing translation for 'onlineSavingsMsg'
Learn More

Palmitic acid, sodium salt, 98%

Catalog No. AC416701000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416701000 100 g Glass bottle
AC416700050 5 g Glass bottle
AC416700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC416701000 Supplier Thermo Scientific Chemicals Supplier No. 416701000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 408-35-5 | C16H31NaO2 | 278.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 408-35-5
Molecular Formula C16H31NaO2
Molecular Weight (g/mol) 278.4
MDL Number MFCD00002749
InChI Key GGXKEBACDBNFAF-UHFFFAOYSA-M
Synonym sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1
PubChem CID 2735111
IUPAC Name sodium;hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)[O-].[Na+]

Specifications

Color White
Loss on Drying 2% max. (110°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)14COONa
Quantity 100 g
Beilstein 02,III,94
Formula Weight 278.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Palmitic acid, sodium salt
Show More Show Less
EINECSNumber 206-988-1

RUO – Research Use Only