missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid Hydrazide 98.0+%, TCI America™

Catalog No. P000425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P000425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P000425G Supplier TCI America Supplier No. P000425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2619-88-7
Molecular Formula C16H34N2O
Molecular Weight (g/mol) 270.461
MDL Number MFCD00066357
InChI Key SSVSELJXJJCANX-UHFFFAOYSA-N
Synonym palmitohydrazide, palmitic acid hydrazide, hexadecanoyl hydrazide, hexadecanoic acid hydrazide, palmiticacidhydrazide, hexadecanohydrazide #, acmc-1cbma, 16-hexadecanoyl hydrazide, hexadecanoic acid,hydrazide, hexdecanoic acid hydrazide
PubChem CID 225536
IUPAC Name hexadecanehydrazide
SMILES CCCCCCCCCCCCCCCC(=O)NN

Specifications

Melting Point 111°C
Color White
Quantity 25 g
Formula Weight 270.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Palmitic Acid Hydrazide