missing translation for 'onlineSavingsMsg'
Learn More

Palmitic acid, 95%

CAS: 57-10-3 | C16H32O2 | 256.43 g/mol

Supplier:  Thermo Scientific Chemicals B203220E

Quantity 2500 g

 View more versions of this product

Catalog No. AAB203220E


Only null left

Description

Description

A saturated fatty acid. Palmitic acid is a saturated fatty acid used in hair care, cosmetics, soaps, paint, rubber, food, pharmaceuticals, animal feed and textiles. It is used to prepare sodium palmitate which is a natural additive in organic products. It is involved in the preparation of cetyl alcohol utilized in the preparation of detergents and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A saturated fatty acid. Palmitic acid is a saturated fatty acid used in hair care, cosmetics, soaps, paint, rubber, food, pharmaceuticals, animal feed and textiles. It is used to prepare sodium palmitate which is a natural additive in organic products. It is involved in the preparation of cetyl alcohol utilized in the preparation of detergents and cosmetics.

Solubility
Soluble in hot alcohol, acetone, benzene, ethyl ether, amyl acetate, propyl alcohol and chloroform. Slightly soluble in cold alcohol and petroleum ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and bases.
Specifications

Chemical Identifiers

57-10-3
256.43
IPCSVZSSVZVIGE-UHFFFAOYSA-N
985
hexadecanoic acid
C16H32O2
MFCD00002747
palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
CHEBI:15756
CCCCCCCCCCCCCCCC(O)=O

Specifications

Palmitic acid
62°C to 65°C
215°C (15 mmHg)
Odorless
1.4273
2500 g
607489
palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
IPCSVZSSVZVIGE-UHFFFAOYSA-N
hexadecanoic acid
985
256.43
57-10-3
0.853
206°C (402°F)
C16H32O2
CH3(CH2)14COOH
MFCD00002747
14,6996
Soluble in hot alcohol,acetone,benzene,ethyl ether,amyl acetate,propyl alcohol and chloroform. Slightly soluble in cold alcohol and petroleum ether. Insoluble in water.
CCCCCCCCCCCCCCCC(O)=O
256.43
CHEBI:15756
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-312-9

RTECSNumber : RT4550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions