missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitoleic acid, >99%, MP Biomedicals™
$1458.77 - $1458.77
Chemical Identifiers
| CAS | 61-25-6 |
|---|---|
| Molecular Formula | C20H22ClNO4 |
| Molecular Weight (g/mol) | 375.849 |
| InChI Key | UOTMYNBWXDUBNX-UHFFFAOYSA-N |
| Synonym | papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan |
| PubChem CID | 6084 |
| IUPAC Name | 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride |
| SMILES | COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15604491
|
MP Biomedicals Inc
0215604491 |
1 g |
Each for $1,458.77
|
|
|||||
Chemical Identifiers
| 61-25-6 | |
| 375.849 | |
| papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan | |
| 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride |
| C20H22ClNO4 | |
| UOTMYNBWXDUBNX-UHFFFAOYSA-N | |
| 6084 | |
| COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl |
Specifications
| 61-25-6 | |
| 0.895g/mL at 20°C | |
| C20H22ClNO4 | |
| papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan | |
| COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl | |
| 375.849 | |
| 254.4 | |
| Frozen Liquid |
| Colorless | |
| ≥99% | |
| 1 g | |
| UOTMYNBWXDUBNX-UHFFFAOYSA-N | |
| 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride | |
| 6084 | |
| >99% | |
| Palmitoleic Acid |
Safety and Handling
EINECSNumber : 206-765-9
Recommended Storage : Store at -0°C.