missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitanilide 98.0+%, TCI America™
Supplier: TCI America P0844500G
Specifications
| Palmitanilide | |
| 90°C | |
| 284°C | |
| MFCD00059287 | |
| N-Palmitoylaniline, N-Phenylpalmitamide | |
| CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1 | |
| 331.544 | |
| 331.54 | |
| Crystalline Powder |
| 6832-98-0 | |
| White | |
| C22H37NO | |
| 500 g | |
| VENJCACPSJGPCM-UHFFFAOYSA-N | |
| N-phenylhexadecanamide | |
| 96171 | |
| ≥98.0% (N) |
Chemical Identifiers
| 6832-98-0 | |
| 331.544 | |
| VENJCACPSJGPCM-UHFFFAOYSA-N | |
| 96171 | |
| CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1 |
| C22H37NO | |
| MFCD00059287 | |
| N-Palmitoylaniline, N-Phenylpalmitamide | |
| N-phenylhexadecanamide |
Safety and Handling
EINECSNumber : (3)-2698
TSCA : No