missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitanilide 98.0+%, TCI America™
$73.16 - $746.97
Chemical Identifiers
| CAS | 6832-98-0 |
|---|---|
| Molecular Formula | C22H37NO |
| Molecular Weight (g/mol) | 331.544 |
| MDL Number | MFCD00059287 |
| InChI Key | VENJCACPSJGPCM-UHFFFAOYSA-N |
| Synonym | N-Palmitoylaniline, N-Phenylpalmitamide |
| PubChem CID | 96171 |
| IUPAC Name | N-phenylhexadecanamide |
| SMILES | CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1 |
Chemical Identifiers
| 6832-98-0 | |
| 331.544 | |
| VENJCACPSJGPCM-UHFFFAOYSA-N | |
| 96171 | |
| CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1 |
| C22H37NO | |
| MFCD00059287 | |
| N-Palmitoylaniline, N-Phenylpalmitamide | |
| N-phenylhexadecanamide |
Specifications
| 6832-98-0 | |
| White | |
| C22H37NO | |
| 25 g | |
| VENJCACPSJGPCM-UHFFFAOYSA-N | |
| N-phenylhexadecanamide | |
| 96171 | |
| ≥98.0% (N) | |
| Palmitanilide |
| 90°C | |
| 284°C | |
| MFCD00059287 | |
| N-Palmitoylaniline, N-Phenylpalmitamide | |
| CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1 | |
| 331.544 | |
| 331.54 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2698
TSCA : No