Learn More
Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL
Pd in 20% HCl
$268.33 - $268.33
Description
Palladium, plasma standard solution finds application as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| Pd in 20% HCl | |
| MFCD03613603 |
Specifications
| Liquid | |
| Pd 1000 μg/mL | |
| UN1789 | |
| 100 mL |
| Pd in 20% HCl | |
| MFCD03613603 | |
| Miscible with water | |
| Palladium, plasma standard solution, Specpure™ |
Safety and Handling
GHS H Statement
H314-H335;
Causes severe skin burns and eye damage.
May cause respiratory irritation.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber : 232-595-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only