missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) nitrate, solution, Pd 10% w/w (cont. Pd)

Catalog No. p-7049057
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12621NH 1 g
AA12621NB 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12621NH Supplier Thermo Scientific Chemicals Supplier No. 012621NH

CAS: 10102-05-3 | N2O6Pd | 230.43 g/mol

Palladium(II) nitrate is used as a reactant for doping activated carbon for catalysis. It acts as a catalyst for hydrogenation of 1-heptyne and naphthalene. It serves as an important raw material for the preparation of platinum-palladium carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cells and in the synthesis of copper palladium alloy thin films. Furthermore, it is used in the synthesis of di-phenyl sulfide-modified palladium-titanium dioxide catalysts for acetylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10102-05-3
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate
PubChem CID 24932
IUPAC Name palladium(2+);dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Brown to Orange
UN Number UN3264
Concentration Pd 10% w/w (cont. Pd)
Solubility Information Miscible with water and dilute nitric acid.
Formula Weight 230.43
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Palladium(II) nitrate, Solution
Hazard Category H290-H314-H332
Hazard Statement GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-265-8
RTECSNumber QV0560000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only