missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Bromide 98.0+%, TCI America™

Catalog No. P19372G
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19372G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19372G Supplier TCI America Supplier No. P19372G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Palladium (II) bromide reagent suitable for laboratory research purpose

Chemical Identifiers

CAS 13444-94-5
Molecular Formula Br2Pd
Molecular Weight (g/mol) 266.23
MDL Number MFCD00011170
InChI Key INIOZDBICVTGEO-UHFFFAOYSA-L
Synonym palladium ii bromide, palladium bromide, palladium bromide pdbr2, palladous bromide, palladium ii bromide, premion, palladium 2+ dibromide, pdbr2, palladiumbromide, palladiumbromide pdbr2
PubChem CID 83469
IUPAC Name palladium(2+) dibromide
SMILES [Br-].[Br-].[Pd++]

Specifications

Color Black-Gray
Physical Form Crystalline Powder
Quantity 2 g
Formula Weight 266.23
Percent Purity ≥98.0% (T)
Chemical Name or Material Palladium(II) Bromide