missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Acetate Trimer 98.0+%, TCI America™

Catalog No. p-7135409
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21061G Supplier TCI America Supplier No. P21061G
Only null left

Chemical Identifiers

CAS 53189-26-7
Molecular Formula C12H24O12Pd3
Molecular Weight (g/mol) 679.572
InChI Key FUKTVTHLBVPABW-UHFFFAOYSA-N
Synonym Hexakis(acetato)tripalladium(II), Tripalladium(II) Hexaacetate
PubChem CID 85470766
IUPAC Name acetic acid;palladium
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pd].[Pd].[Pd]

Specifications

Quantity 1 g
Formula Weight 673.52
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Palladium(II) Acetate Trimer
EINECSNumber (2)-0693
TSCA No