accessibility menu, dialog, popup

UserName

CAS RN 53189-26-7

CH3OHOCH3OHOCH3OHOCH3OHOCH3OHOCH3OHOPdPdPd

CAS RN 53189-26-7

IUPAC Name: hexakis(acetic acid) tripalladium
Synonyms: hexakis(acetic acid) tripalladium
Molecular Weight (g/mol): 679.57
Molecular Formula: C12H24O12Pd3
InChi Key: FUKTVTHLBVPABW-UHFFFAOYSA-N
SMILES: [Pd].[Pd].[Pd].CC(O)=O.CC(O)=O.CC(O)=O.CC(O)=O.CC(O)=O.CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results