missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Acetate 98.0+%, TCI America™

Supplier:  TCI America A14245G

 View more versions of this product

Catalog No. A14245G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Palladium(II) Acetate
Crystalline Powder
C4H6O4Pd
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
[Pd++].CC([O-])=O.CC([O-])=O
224.51
224.51
3375-31-3
5 g
MFCD00012453
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium(2+) diacetate
167845
≥98.0% (T)

Chemical Identifiers

3375-31-3
224.51
YJVFFLUZDVXJQI-UHFFFAOYSA-L
167845
[Pd++].CC([O-])=O.CC([O-])=O
C4H6O4Pd
MFCD00012453
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
palladium(2+) diacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0693

RTECSNumber : AJ1900000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions