missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Acetate 98.0+%, TCI America™

Catalog No. p-7117427
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14241G 1 g
A14245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14241G Supplier TCI America Supplier No. A14241G
Only null left

Chemical Identifiers

CAS 3375-31-3
Molecular Formula C4H6O4Pd
Molecular Weight (g/mol) 224.51
MDL Number MFCD00012453
InChI Key YJVFFLUZDVXJQI-UHFFFAOYSA-L
Synonym palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
PubChem CID 167845
IUPAC Name palladium(2+) diacetate
SMILES [Pd++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 224.51
Percent Purity ≥98.0% (T)
Chemical Name or Material Palladium(II) Acetate
EINECSNumber (2)-0693
RTECSNumber AJ1900000
TSCA Yes