missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) Acetate 98.0+%, TCI America™
Supplier: TCI America A14241G
Specifications
| Palladium(II) Acetate | |
| Crystalline Powder | |
| C4H6O4Pd | |
| palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
| [Pd++].CC([O-])=O.CC([O-])=O | |
| 224.51 | |
| 224.51 |
| 3375-31-3 | |
| 1 g | |
| MFCD00012453 | |
| YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
| palladium(2+) diacetate | |
| 167845 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3375-31-3 | |
| 224.51 | |
| YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
| 167845 | |
| [Pd++].CC([O-])=O.CC([O-])=O |
| C4H6O4Pd | |
| MFCD00012453 | |
| palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
| palladium(2+) diacetate |
Safety and Handling
EINECSNumber : (2)-0693
RTECSNumber : AJ1900000
TSCA : Yes