missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) acetate (47% Pd), MilliporeSigma™

For synthesis

Supplier:  Technidata 8180568500

 View more versions of this product

Catalog No. M8180568500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Palladium(II) acetate (47% Pd)
500 mg
MFCD00012453
Soluble in water at 20°C
[Pd++].CC([O-])=O.CC([O-])=O
224.51
224.5
Glass Bottle
3375-31-3
C4H6O4Pd
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium(2+) diacetate
167845
Slight

Chemical Identifiers

3375-31-3
224.51
YJVFFLUZDVXJQI-UHFFFAOYSA-L
167845
[Pd++].CC([O-])=O.CC([O-])=O
C4H6O4Pd
MFCD00012453
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
palladium(2+) diacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H318: Causes serious eye damage.

EINECSNumber : 222-164-4

SDS
Documents

Documents

Promotions

Promotions